missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35340-100ul
This item is not returnable.
View return policy
Description
NRBF2 Polyclonal antibody specifically detects NRBF2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| NRBF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-220 of human NRBF2 (NP_110386.2).,, Sequence:, LAHQQSRRADRLLAAGKYEEAISCHKKAAAYLSEAMKLTQSEQAHLSLELQRDSHMKQLLLIQERWKRAQREERLKAQQNTDKDAAAHLQTSHKPSAEDAEGQSPLSQKYSPSTEKCLPEIQGIFDRDPDTLLYLLQQKSEPAEPCIGSKAPKDDKTIIEEQATKIADLKRHVEFLVAENERLRKENKQLKAEKARLLKGPIEKELDVDAD | |
| 100 μL | |
| Signal Transduction | |
| 29982 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction