missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19583
This item is not returnable.
View return policy
Description
NRBF2 Polyclonal specifically detects NRBF2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| NRBF2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2 | |
| Rabbit | |
| 32 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 29982 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96F24 | |
| NRBF2 | |
| Recombinant protein encompassing a sequence within the center region of human NRBF2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Chicken (81%), Rat (92%), Bovine (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction