Learn More
NRBP1 Mouse anti-Canine, Human, Rat, Clone: OTI7E2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500451
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.Specifications
| NRBP1 | |
| Monoclonal | |
| Unconjugated | |
| NRBP1 | |
| BCON3, MADM, MUDPNP, NRBP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29959, 475704, 619579 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| OTI7E2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UHY1 | |
| NRBP1 | |
| Full-length protein expressed in 293T cell transfected with human NRBP1 expression vector | |
| 100 μg | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.