missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBP2 Antibody (2D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00340371-M03
This item is not returnable.
View return policy
Description
NRBP2 Monoclonal antibody specifically detects NRBP2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| NRBP2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp434P086, MGC138699, nuclear receptor binding protein 2, nuclear receptor-binding protein 2, pp9320, Transformation-related gene 16 protein, transformation-related protein 16, TRG16, TRG-16 | |
| NRBP2 (NP_848659, 151 a.a. ∽ 258 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PRVLAPPPEEVQKAKTPTPEPFDSETRKVIQMQCNLERSEDKARWHLTLLLVLEDRLHRQLTYDLLPTDSAQDLASELVHYGFLHEDDRMKLAAFLESTFLKYRGTQA | |
| 0.1 mg | |
| Protein Kinase | |
| 340371 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 2D5 | |
| Western Blot 1:500, ELISA | |
| NP_848659 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction