missing translation for 'onlineSavingsMsg'
Learn More

NRBP2 Mouse anti-Human, CoraLite 594, Clone: 4F10H7, Proteintech

Product Code. 16811066 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16811066 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16811066 Supplier Proteintech Supplier No. CL59466687100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NRBP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NRBP2. This antibody recognizes human, mouse, rat, pig antigen. The NRBP2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.

Specifications

Antigen NRBP2
Applications Immunofluorescence
Classification Monoclonal
Clone 4F10H7
Conjugate CoraLite 594
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 340371
Target Species Human, Mouse, Rat, Pig
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.