missing translation for 'onlineSavingsMsg'
Learn More

NRC-2694, MedChemExpress

Product Code. 16481889
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16481889

Brand: Medchem Express HY19909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NRC-2694 is an epidermal growth factor receptor (EGFR) antagonist with anti-cancer and anti-proliferative properties.

NRC-2694 is an epidermal growth factor receptor (EGFR) antagonist with anti-cancer and anti-proliferative properties.

Specifications

Chemical Name or Material NRC-2694
CAS 936446-61-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26N4O3
Quantity 1 mg
Solubility Information DMSO : 250 mg/mL (597.39 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=CC(C#C)=C3)=C2C=C1OCCCN4CCOCC4
Molecular Weight (g/mol) 418.49
Formula Weight 418.49
Percent Purity 98.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.