Learn More
Invitrogen™ NRCAM Monoclonal Antibody (N364/51), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545683
Product Code. 17866692
  • € 455.00 / 100µg
Description
Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ~50% identity with Neurofascin. 1 μg/mL of MA5-45683 was sufficient for detection of NrCAM in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. This antibody was formerly sold as clone S364-51.
Neuron-glia-related cell adhesion molecule (NRCAM) is a gene located on chromosome 7q31.1, encoding a protein that belongs to the immunoglobulin superfamily of cell adhesion molecules. NRCAM is expressed in the nervous system and plays a vital role in the development and maintenance of neural connections. This adhesion molecule facilitates neuron-neuron and neuron-glia interactions, impacting processes such as axon guidance, synapse formation, and cell signaling, which are crucial for neural development and plasticity. NRCAM is involved in the modulation of signal transduction pathways that influence neuronal growth and repair, contributing to the stabilization of synaptic structures. Alterations in NRCAM expression or function have been linked to neurodevelopmental disorders, including autism spectrum disorders, and certain psychiatric conditions, reflecting its importance in maintaining neural circuit integrity. Research into NRCAM continues to explore its therapeutic potential in neurobiology and its implications in neurological and psychiatric disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NRCAM
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 2mM EDTA and no preservative; pH 7.4
P97686, Q810U4, Q92823
NRCAM
Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot, Immunocytochemistry
N364/51
PerCP
NRCAM
ankyrin-binding cell adhesion molecule NrCAM; Bravo; C030017F07Rik; C130076O07Rik; hBravo; KIAA0343; mBravo; MGC138845; MGC138846; mKIAA0343; neuronal cell adhesion molecule; Neuronal surface protein Bravo; neuron-glia-CAM-related cell adhesion molecule; ng-CAM-related; NgCAM-related cell adhesion molecule; Nr CAM; Nrcam; nr-CAM; rBravo
Mouse
Protein G
RUO
319504, 4897, 497815
4°C