missing translation for 'onlineSavingsMsg'
Learn More

Nrcam Mouse anti-Human, Mouse, Rat, ATTO 488, Clone:S364-51, Abnova™

Product Code. 16250020
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16250020 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16250020 Supplier Abnova Supplier No. MAB17670.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant mouse Nrcam.

Specifications

Antigen Nrcam
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S364-51
Conjugate Atto 488
Description neuron-glia-CAM-related cell adhesion molecule
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Nrcam
Gene Alias Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 30-845 at extracellular domain of mouse Nrcam.
Quantity 100 μg
Gene ID (Entrez) 319504
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.