missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRCAM Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68291-1-IG-20UL
This item is not returnable.
View return policy
Description
The NRCAM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NRCAM. This antibody recognizes human, mouse, rat, pig antigen. The NRCAM antibody has been validated for the following applications: WB, IF-P, ELISA analysis.Specifications
| NRCAM | |
| Monoclonal | |
| 1000 μg/mL | |
| KIAA0343; MGC138845; MGC138846 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 130-140 kDa | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 2E1C3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 4897 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction