missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRCAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116925
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y, Mouse spinal cord, Mouse brain Immunogen sequence: QMISALEVPL DPKLLEDLVQ PPTITQQSPK DYIIDPRENI VIQCEAKGKP PPSFSWTRNG THFDIDKDPL VTMKPGTGTL IINIMSEGKA E.
Neuron-glia-related cell adhesion molecule (NRCAM) is a gene located on chromosome 7q31.1, encoding a protein that belongs to the immunoglobulin superfamily of cell adhesion molecules. NRCAM is expressed in the nervous system and plays a vital role in the development and maintenance of neural connections. This adhesion molecule facilitates neuron-neuron and neuron-glia interactions, impacting processes such as axon guidance, synapse formation, and cell signaling, which are crucial for neural development and plasticity. NRCAM is involved in the modulation of signal transduction pathways that influence neuronal growth and repair, contributing to the stabilization of synaptic structures. Alterations in NRCAM expression or function have been linked to neurodevelopmental disorders, including autism spectrum disorders, and certain psychiatric conditions, reflecting its importance in maintaining neural circuit integrity. Research into NRCAM continues to explore its therapeutic potential in neurobiology and its implications in neurological and psychiatric disorders.
Specifications
| NRCAM | |
| Polyclonal | |
| Unconjugated | |
| NRCAM | |
| ankyrin-binding cell adhesion molecule NrCAM; Bravo; C030017F07Rik; C130076O07Rik; hBravo; KIAA0343; mBravo; MGC138845; MGC138846; mKIAA0343; neuronal cell adhesion molecule; Neuronal surface protein Bravo; neuron-glia-CAM-related cell adhesion molecule; ng-CAM-related; NgCAM-related cell adhesion molecule; Nr CAM; Nrcam; nr-CAM; rBravo | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 319504, 4897 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q810U4, Q92823 | |
| NRCAM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-115 of human NRCAM (Q92823). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction