missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRDE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562218
This item is not returnable.
View return policy
Description
Immunogen sequence: PNNTALWQKY LLFCQSQFST FSISKIHSLY GKCLSTLSAV KDGSILSHPA LPGTEEAMFA LFLQQCHFLR QAGHSEKAIS LFQAMVDFTF FKPDSVKDLP TKGQVEFFEP FWDSGEPRAG EKGARGWKAW MHQQERGGWV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
NRDE2 gene ontology (GO) annotations include RNA interference; chromatin silencing by small RNA.
Specifications
| NRDE2 | |
| Polyclonal | |
| Unconjugated | |
| NRDE2 | |
| 6720454P05Rik; C14orf102; Nrde2; nrde-2 necessary for RNA interference, domain containing; NRDE-2, necessary for RNA interference, domain containing; Nuclear exosome regulator NRDE2; Protein NRDE2 homolog; UPF0614 protein C14orf102; UPF0614 protein C14orf102 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55051 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7Z3 | |
| NRDE2 | |
| Recombinant protein corresponding to Human NRDE2. Recombinant protein control fragment (Product #RP-89044). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction