missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nrf1 Antibody (NRF1/2609), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08773DL594
This item is not returnable.
View return policy
Description
Nrf1 Monoclonal antibody specifically detects Nrf1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Nrf1 | |
| Monoclonal | |
| DyLight 594 | |
| Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1 | |
| Recombinant full-length human Nrf1 protein (Uniprot: Q16656) | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 4899 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| NRF1/2609 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction