missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551088
This item is not returnable.
View return policy
Description
Neuregulin-2 Polyclonal Antibody detects endogenous levels of Neuregulin-2 protein.
Neuregulin 2 is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1, another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described.
Specifications
| NRG2 | |
| Polyclonal | |
| Unconjugated | |
| NRG2 | |
| divergent of neuregulin-1; Don1; DON-1; HRG2; neural- and thymus-derived activator for ErbB kinases; neuregulin 2; Neuregulin-2; NRG2; NRG-2; NTAK; NTAK alpha1; NTAK_alpha2a; pro-neuregulin-2, membrane-bound isoform; Pro-NRG2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100042150, 432361, 9542 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| O14511, O35569, P56974 | |
| NRG2 | |
| Synthesized peptide derived from the Internal region of human Neuregulin-2 (aa 361-410). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction