missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRG4 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00145957-B01P
This item is not returnable.
View return policy
Description
NRG4 Polyclonal antibody specifically detects NRG4 in Human samples. It is validated for Western Blot
Specifications
| NRG4 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4 | |
| NRG4 (NP_612640.1, 1 a.a. - 115 a.a.) full-length human protein. MPTDHEEPCGPSHKSFCLNGGLCYVIPTIPSPFCRCVENYTGARCEEVFLPGSSIQTKSNLFEAFVALAVLVTLIIGAFYFLCRKGHFQRASSVQYDINLVETSSTSAHHSHEQH | |
| 0.05 mg | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 145957 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_612640.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction