missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558281
This item is not returnable.
View return policy
Description
Immunogen sequence: MDCCTENACS KPDDDILDIP LDDPGAN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.
Specifications
| NRGN | |
| Polyclonal | |
| Unconjugated | |
| NRGN | |
| 0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; calmodulin-binding protein; hng; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); ng; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3; similar to neurogranin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92686 | |
| NRGN | |
| Recombinant protein corresponding to Human NRGN. Recombinant protein control fragment (Product #RP-97413). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction