missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRIP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-33208
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NRIP Polyclonal specifically detects NRIP in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.
Especificaciones
| NRIP | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS with No Preservative | |
| 1200006M05Rik, Androgen receptor complex-associated protein, ARCAP, DDB1 and CUL4 associated factor 6, DDB1- and CUL4-associated factor 6, FLJ23798, IQ motif and WD repeat-containing protein 1, IQ motif and WD repeats 1, IQWD1, MSTP055, NRIP, Nuclear receptor interaction protein, PC326 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55827 | |
| Human, Mouse, Rat, Canine | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Q58WW2 | |
| DCAF6 | |
| A portion of amino acids 800-850 of human NRIP was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used is 100% homologous in human (isoforms a and b), mouse, rat and dog, 88% homologous in zebrafish and 82% homologous in xenopus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido