missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRIP3 Antibody (OTI1G6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73065F
This item is not returnable.
View return policy
Description
NRIP3 Monoclonal specifically detects NRIP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| NRIP3 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NRIP3 | |
| Full length recombinant protein of human NRIP3 (NP_065696) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1G6 | |
| Flow Cytometry | |
| C11orf14, chromosome 11 open reading frame 14, nuclear receptor interacting protein 3, nuclear receptor-interacting protein 3, NY-SAR-105, Sarcoma antigen NY-SAR-105 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 56675.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction