missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRK Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98072IR
This item is not returnable.
View return policy
Description
NRK Polyclonal specifically detects NRK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NRK | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| DKFZp686A17109, EC 2.7.11, EC 2.7.11.1, FLJ16788, MGC131849, NESK, Nik related kinase, nik-related protein kinase | |
| Produced in rabbits immunized with E. coli-derived Human NRK fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 203447 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction