missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRK Polyclonal Antibody

Description
Predicted to react with Mouse samples.
The mouse ortholog of this gene encodes a protein kinase required for JNK activation. The encoded protein may be involved in the induction of actin polymerization in late embryogenesis.
Specifications
Specifications
| Antigen | NRK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NRK |
| Gene Accession No. | Q7Z2Y5, Q9R0G8 |
| Gene Alias | AA407323; AA409510; AI225895; AV168563; AW538185; Nck-interacting kinase-like embryo specific kinase; Nesk; Nik related kinase; NIK-like embryo-specific kinase; nik-related protein kinase; Nrk |
| Gene Symbols | NRK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?