missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSF Antibody (CL0381), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42048
This item is not returnable.
View return policy
Description
NRSF Monoclonal antibody specifically detects NRSF in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NRSF | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL0381 | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SPPLPKENLREEASGDQKLLNTGEGNKEAPLQKVGAEEADESLPGLAANINESTHISSSGQNLNTPEGETLNGKHQTDSIVCEMKMDTDQNTRENLTGINSTVEEPVSPMLPPSAVEEREAVSKTALA | |
| 0.1 mL | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| 5978 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction