missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSF Antibody (SR1230), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | NRSF |
|---|---|
| Clone | SR1230 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681394
|
Novus Biologicals
NBP3-21822-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642674
|
Novus Biologicals
NBP3-21822-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NRSF Monoclonal antibody specifically detects NRSF in Human samples. It is validated for Western BlotSpecifications
| NRSF | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| A synthesized peptide derived from human NRSF (Uniprot #: Q13127) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1230 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5978 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title