missing translation for 'onlineSavingsMsg'
Learn More

NRXN1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-00219

Product Code. 18208970

  • € 543.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NRXN1 Polyclonal specifically detects NRXN1 in Human samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NRXN1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
DKFZp313P2036, FLJ35941, KIAA0578Neurexin I-alpha, neurexin 1, neurexin I, Neurexin I-beta, neurexin-1-alpha, neurexin-1-beta, PTHSL2
Goat
Affinity Purified
RUO
9378
Human
IgG
Western Blot, Flow Cytometry, ELISA
0.5 mg/mL
Western Blot 0.5 - 1 μg/mL, Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:128000
Q9ULB1
NRXN1
Peptide with sequence C-KEKQPSSAKSSNKN corresponding to C-Terminus according to NP_620072.1.
0.1 mg
Primary
This antibody is expected to recognize both reported isoforms (NP_004792.1 and NP_620072.1).
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only