missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 3728

Description
NS 3728 is a volume-regulated anion channel (VRAC) blocker (IC50 = 460 nM). Blocks red blood cell chloride conductance (IC50 = 0.6 μM). NS 3728 also inhibits Ca2+-activated Cl- current (CaCC) by anoctamin (ANO) in an apparently voltage-dependent manner (IC50 against ANO1 = 2.1 and 0.7 μM at -95 and +50 mV, respectively).
Specifications
Specifications
| Chemical Name or Material | N-[3,5-bis(Trifluoromethyl)phenyl]-N'-[4-bromo-2-(2 H-tetrazol-5-yl)phenyl]urea |
| CAS | 265646-85-3 |
| Quantity | 50 mg |
| Target | Other Chloride Channel Blockers |
| Molecular Formula | C16H9BrF6N6O |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?