missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 383

Description
NS 383 is an ASIC blocker (IC50 values are 0.44, 2.1 μM and no effect at rat ASIC1a, ASIC3 and ASIC2a, respectively: IC50 value = 0.12 μM at human ASIC1a with no effect at ASIC2a or ASIC3). Inhibition was also observed at heteromeric ASIC channels (IC50 values are 0.79, 0.87 and 4.5 uM at rat ASIC1a+3, ASIC1a+2a and ASIC2a+3; IC5O values are 0.33 and 0.69 uM at human ASIC1a+2a and ASIC1a+3, with no effect at ASIC2a+3). Attenuates pathophysiological nociceptive behaviors in CFA-inflamed and CCI rats.
Specifications
Specifications
| Chemical Name or Material | 8-Ethyl-6,7,8,9-tetrahydro-5-phenyl-1 H-pyrrolo[3,2-h]isoquinoline-2,3-dione-3-oxime |
| CAS | 309711-59-9 |
| Quantity | 10 mg |
| Target | Acid-sensing Ion Channel Blockers |
| Molecular Formula | C19H19N3O2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?