Learn More
Tocris Bioscience™ NS 8593 Hydrochloride
GREENER_CHOICE
Selective negative modulator of KCa2 (SK) channels; inhibits SK currents
Brand:  Tocris Bioscience™ 4597/10
Encompass
Product Code. 15756206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.
Specifications
Specifications
NS 8593 hydrochloride
N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride
Soluble to 100mM in DMSO and to 20mM in ethanol
Bioactive Small Molecule
875755-24-1
10 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use