Learn More
NSBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23955-1-AP-150UL
 View more versions of this product
Product Code. 16812555
  • € 410.00 / 150µL
Description
This gene encodes a nuclear protein with similarities to the high mobility group proteins, HMG14 and HMG17, which suggests that this protein may function as a nucleosomal binding and transcriptional activating protein.
Specifications
Specifications
NSBP1
Polyclonal
Unconjugated
HMGN5
HMGN5, NSBP1, nucleosomal binding protein 1, Nucleosome binding protein 1
Rabbit
Antigen Affinity Chromatography
RUO
79366
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P82970
HMGN5
NSBP1 Fusion Protein Ag20884
150 μL