missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 540.75
Specifications
| Antigen | NSD3 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452692
|
Novus Biologicals
NBP1-89851 |
0.1 mL |
€ 572.00 € 540.75 / 0.10mL Save € 31.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18466001
|
Novus Biologicals
NBP1-89851-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSD3 Polyclonal antibody specifically detects NSD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| NSD3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.2) and 40% Glycerol | |
| 54904 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NTQPEQTNAGEVASSLSSTEIRRHSQRRHTSAEEEEPPPVKIAWKTAAARKSLPASITMHKGSLDLQKCNMSPVVKIEQVFALQNATGDGKFIDQFVYSTKGIGNKTEISVRGQDRLIISTPNQRNEKPTQSVSSPEATS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title