missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSDHL Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99165AF594
This item is not returnable.
View return policy
Description
NSDHL Polyclonal antibody specifically detects NSDHL in Human samples. It is validated for Western Blot
Specifications
| NSDHL | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| EC 1.1.1.170, H105e3, member 1, NAD(P) dependent steroid dehydrogenase-like, SDR31E1, sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating, XAP104 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NSDHL | |
| 0.1 mL | |
| metabolism | |
| 50814 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction