missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nse2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76263G
This item is not returnable.
View return policy
Description
Nse2 Polyclonal antibody specifically detects Nse2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Nse2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 286053 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 488 | |
| C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human MMS21. The immunogen is located within amino acids 170 - 220 of MMS21. Amino Acid Squence: CIVRMIESRQKRKKKA | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction