missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSFL1C Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36529FR
This item is not returnable.
View return policy
Description
NSFL1C Polyclonal antibody specifically detects NSFL1C in Human, Mouse samples. It is validated for Western Blot
Specifications
| NSFL1C | |
| Polyclonal | |
| Western Blot | |
| dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, p47, p97 cofactor p47, SHP1 homolog, UBX domain protein 2C, UBX domain-containing protein 2C, UBX1, UBXD10, UBXN2CMGC3347 | |
| The immunogen recognized by the antibody maps to a region between residue 25 to 75 of human NSFL1 (p97) Cofactor (p47) using the numbering given in entry NP_057227.2. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55968 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction