Learn More
NSFL1C Mouse, anti-Human, CoraLite Plus 555, Clone: 1F12E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-68182-20UL
Description
The NSFL1C antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NSFL1C. This antibody recognizes human, mouse, rat, pig antigen. The NSFL1C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 555.
Specifications
| NSFL1C | |
| 1F12E12 | |
| CoraLite Plus 555 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, NSFL1C, p47, p97 cofactor p47, UBX1, UBXD10, UBXN2C | |
| 20 μL | |
| 55968 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |