Learn More
Invitrogen™ NSG2 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5143289
Product Code. 17829082
  • € 465.00 / 100µL
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
NSG2 is a protein coding gene. Gene ontology (GO) annotation include cytoplasmic vesicle membrane; dendrite; early endosome; early endosome membrane; endosome; Golgi apparatus; Golgi cis cisterna membrane; integral component of membrane; late endosome; lysosomal lumen; multivesicular body membrane; trans-Golgi network membrane.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NSG2
Polyclonal
Unconjugated
NSG2
CALY3; HMP19; HMP19 protein; hypothalamus golgi apparatus expressed 19 kDa protein; neuron specific gene family member 2; Neuronal vesicle trafficking-associated protein 2; neuron-specific protein family member 2; NSG2; p19 protein; protein 8.5; Protein p19
Goat
Ammonium Sulfate Precipitation
RUO
18197, 497878
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P47759
NSG2
Peptide with sequence C-HEPKPPKTQGH (aa 161-171).
100 μg
Primary
Mouse, Rat