missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NSMAF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | NSMAF |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066884
|
Novus Biologicals™
NBP3-30930-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067115
|
Novus Biologicals™
NBP3-30930-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSMAF Polyclonal antibody specifically detects NSMAF in Human samples. It is validated for Western BlotSpecifications
| NSMAF | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Immunology, Innate Immunity, Signal Transduction | |
| Phosphate buffered saline | |
| 8439 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Factor associated with neutral sphingomyelinase activation, FANFactor associated with N-SMase activation, neutral sphingomyelinase (N-SMase) activation associated factor, protein FAN | |
| Synthetic peptide derived from human NSMAF protein. | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title