missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSMCE1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98533MFV500
This item is not returnable.
View return policy
Description
NSMCE1 Polyclonal antibody specifically detects NSMCE1 in Human samples. It is validated for Western Blot
Specifications
| NSMCE1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 500 SE | |
| Non-SMC element 1 homolog, non-SMC element 1 homolog (S. cerevisiae), non-structural maintenance of chromosomes element 1 homolog, NSE1 | |
| Produced in rabbits immunized with E. coli-derived Human NSMCE1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 197370 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction