missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSMCE2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00286053-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MPGRSSSNSGSTGFISFSGVESALSSLKNFQACINSGMDTASSVALDLVESQTEVSSEYSMDKAMVEFATLDRQLNHYVKAVQSTINHVKEERPEKIPDLKLLVEKKFLALQSKNSDADFQNNEKFVQFKQQLKELKKQCGLQADREADGTEGVDEDIIVTQSQTNFTCPITKEEMKKPVKNKVCGHTYEEDAIVRMIESRQKRKKKAYCPQIGCSHTDIRKSDLIQDEALRRAIENHNKKRHRHSESpecifications
| NSMCE2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_173685 | |
| NSMCE2 | |
| C8orf36 (NP_775956, 1 a.a. ∼ 247 a.a) full-length human protein. | |
| RUO | |
| 286053 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| non-SMC element 2, MMS21 homolog (S. cerevisiae) | |
| C8orf36/FLJ32440/MMS21/NSE2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction