missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSP 5 alpha 3 alpha Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-518AF532
This item is not returnable.
View return policy
Description
NSP 5 alpha 3 alpha Polyclonal specifically detects NSP 5 alpha 3 alpha in Human samples. It is validated for Western Blot.
Specifications
| NSP 5 alpha 3 alpha | |
| Polyclonal | |
| Western Blot | |
| cytospin-B, CYTSBcytospin B, FLJ36955, HCMOGT-1, NSP, NSP5, Nuclear structure protein 5, sperm antigen HCMOGT 1, Sperm antigen HCMOGT-1, sperm antigen with calponin homology and coiled coil domains 1, sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure protein NSP5b3b | |
| Rabbit | |
| 0.1 mL | |
| 92521 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SPECC1 | |
| An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 200-300). [UniProtNo. Q5M775] | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction