missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSP 5 alpha 3 alpha Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-518V
This item is not returnable.
View return policy
Description
NSP 5 alpha 3 alpha Polyclonal specifically detects NSP 5 alpha 3 alpha in Human samples. It is validated for Western Blot.
Specifications
| NSP 5 alpha 3 alpha | |
| Polyclonal | |
| Western Blot | |
| Q5M775 | |
| SPECC1 | |
| An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 200-300). [UniProt# Q5M775] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen has 93% homology with mouse and 87% homology with rat. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| cytospin-B, CYTSBcytospin B, FLJ36955, HCMOGT-1, NSP, NSP5, Nuclear structure protein 5, sperm antigen HCMOGT 1, Sperm antigen HCMOGT-1, sperm antigen with calponin homology and coiled coil domains 1, sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure protein NSP5b3b | |
| Rabbit | |
| 80 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience | |
| 92521 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction