missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSP5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582761
This item is not returnable.
View return policy
Description
Immunogen sequence: APREFSVTVS RERSVPRGPS NPRKSVSSPT SSNTPTPTKH LRTPSTKPKQ ENEGGEKAAL ESQVRELLAE AKAKDSEINR LRSELKKYKE KRTLNA Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 77%.
The protein encoded by this gene belongs to the cytospin-A family. It is localized in the nucleus, and highly expressed in testis and some cancer cell lines. A chromosomal translocation involving this gene and platelet-derived growth factor receptor, beta gene (PDGFRB) may be a cause of juvenile myelomonocytic leukemia. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| NSP5 | |
| Polyclonal | |
| Unconjugated | |
| Specc1 | |
| 2810012G08Rik; B230396K10Rik; cytokinesis and spindle organization B; cytospin B; cytospin-B; CYTSB; HCMOGT1; HCMOGT-1; NSP; NSP5; NSP5a3a; NSP5alpha3alpha; Nuclear structure protein 5; RGD1309718; SPECC1; spectrin domain with coiled-coils 1; sperm antigen HCMOGT-1; Sperm antigen with calponin homology and coiled-coil domains 1; sperm antigen with calponin-like and coiled coil domains 1; structure protein NSP5a3a; structure protein NSP5a3b; structure protein NSP5b3a; structure protein NSP5b3b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92521 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5M775 | |
| Specc1 | |
| Recombinant protein corresponding to Human SPECC1. Recombinant protein control fragment (Product #RP-93234). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction