missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSRP1 Antibody, FITC, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41139F
This item is not returnable.
View return policy
Description
NSRP1 Polyclonal antibody specifically detects NSRP1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
Specifications
| NSRP1 | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 84081 | |
| Store at 4°C in the dark. | |
| IgY |
| ELISA, Immunofluorescence | |
| FITC | |
| CCDC55, coiled-coil domain containing 55, coiled-coil domain-containing protein 55, DKFZp434K1421, FLJ37233, HSPC095, NSRP70, nuclear speckle splicing regulatory protein 1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human CCDC55. The immunogen is located within amino acids 430 - 480 of CCDC55. Amino Acid Squence: VQSSERNQDRKESSPN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction