missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | NSUN6 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50-1:200 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633480
|
Novus Biologicals
NBP2-93013-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686951
|
Novus Biologicals
NBP2-93013-0.1ml |
0.1 mL |
€ 470.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSUN6 Polyclonal antibody specifically detects NSUN6 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| NSUN6 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| EC 2.1.1, EC 2.1.1.-, EC 2.1.1.52, FLJ23743, NOL1/NOP2/Sun and PUA domain-containing protein 1, NOL1/NOP2/Sun domain family 6, NOL1/NOP2/Sun domain family member 6, NOL1/NOP2/Sun domain family, member 6, NOP2/Sun domain family, member 6,4933414E04Rik, NOPD1, nucleolar protein (NOL1/NOP2/sun) and PUA domains 1, putative methyltransferase NSUN6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-469 of human NSUN6 (NP_872349.1). NLPSALVSHVLNPQPGEKILDLCAAPGGKTTHIAALMHDQGEVIALDKIFNKVEKIKQNALLLGLNSIRAFCFDGTKAVKLDMVEDTEGEPPFLPESFDRILLDAPCSGMGQRPNMACTWSVKEVASYQPLQRKLFTAAVQLLKPEGVLVYSTCTITLAENEEQVAWALTKFPCLQLQPQEPQIGGEGMRGAGLSCEQLKQLQRFDPSAVPLPDTDMDSLREARREDMLRLANKDSIGFFIAKFVKCKST | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 221078 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title