missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT-proBNP Antibody (11D1cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13059
This item is not returnable.
View return policy
Description
NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA.
Specifications
| NT-proBNP | |
| Monoclonal | |
| Unconjugated | |
| P16860 | |
| NPPB | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 28-45, conjugated with carrier protein. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human NT-proBNP and proBNP. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Western Blot, ELISA | |
| 11D1cc | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Mouse | |
| 15 kDa | |
| 0.2 mg | |
| Cytokine Research, Immunology, Neuroscience | |
| 4879 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction