missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT-proBNP Antibody (5B6cc) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-13111JF635
This item is not returnable.
View return policy
Description
NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for ELISA,Western Blot
Specifications
| NT-proBNP | |
| Monoclonal | |
| Janelia Fluor 635 | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 1-12, conjugated with carrier protein. | |
| 0.2 mL | |
| Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| 4879 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot | |
| 5B6cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction