Learn More
NT5C3 Antibody (009), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90252JF646
This item is not returnable.
View return policy
Description
NT5C3 Monoclonal antibody specifically detects NT5C3 in Human samples. It is validated for Western Blot, Immunoprecipitation