missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C3L Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98136FR
This item is not returnable.
View return policy
Description
NT5C3L Polyclonal antibody specifically detects NT5C3L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NT5C3L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 5'-nucleotidase, cytosolic III-like | |
| Produced in rabbits immunized with E. coli-derived Human NT5C3L fragment. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 115024 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction