missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5D1 Monoclonal Antibody (OTI3A6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525210
This item is not returnable.
View return policy
Description
NT5D1 Monoclonal Antibody for Western Blot, IHC (P), Flow
While the exact function of the protein encoded by this gene is not known, it belongs to the 5'(3')-deoxyribonucleotidase family.
Specifications
| NT5D1 | |
| Monoclonal | |
| 0.78 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TFE4 | |
| NT5DC1 | |
| Full length human recombinant protein of NT5DC1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Canine, Monkey | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A6 | |
| Unconjugated | |
| NT5DC1 | |
| 5'-nucleotidase domain containing 1; 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1; 5'-nucleotidase, cytosolic II-like 1 protein; C6orf200; LP2642; Nt5c2l1; NT5DC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 221294, 475036 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction