missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5DC1 Antibody (OTI1E11), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73078C
This item is not returnable.
View return policy
Description
NT5DC1 Monoclonal specifically detects NT5DC1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| NT5DC1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NT5DC1 | |
| Full length recombinant protein of human NT5DC1 (NP_689942) produced in HEK293T cells. | |
| 0.1 mL | |
| 221294.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1E11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 5′-nucleotidase domain containing 1,5′-nucleotidase domain-containing protein 1, 5′-nucleotidase, cytosolic II-like 1, C6orf200, dJ486I3.1, LP2642, MGC131837, MGC24302,5′-nucleotidase, cytosolic II-like 1 protein, NT5C2L1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction