Learn More
NT5DC1 Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501591S
Encompass_Preferred
 View more versions of this product
Product Code. 17574651
  • € 242.00 / 30µL
Description
NT5DC1 Monoclonal antibody specifically detects NT5DC1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
NT5DC1
Monoclonal
0.59 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5TFE4
Mouse
Affinity chromatography
RUO
221294
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2A1
Unconjugated
NT5DC1
5'-nucleotidase domain containing 1; 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1; 5'-nucleotidase, cytosolic II-like 1 protein; C6orf200; LP2642; Nt5c2l1; NT5DC1
Full length human recombinant protein of human NT5DC1 produced in HEK293T cell.