missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5M, Mouse anti-Human, Clone: 3E2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00056953-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: CTSPIKMFKYCPYEKYAWVEKYFGPDFLEQIVLTRDKTVVSADLLIDDRPDITGAEPTPSWEHVLFTACHNQHLQLQPPRRRLHSWADDWKAILDSKRPCSpecifications
| NT5M | |
| Monoclonal | |
| Unconjugated | |
| 5',3'-nucleotidase, mitochondrial | |
| dNT-2/dNT2/mdN | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3E2 | |
| In 1x PBS, pH 7.4 | |
| BC035838 | |
| NT5M | |
| NT5M (AAH35838, 129 a.a. ∼ 228 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 56953 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction