missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5M Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 184.00 - € 485.00
Specifications
| Antigen | NT5M |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17750334
![]() |
Invitrogen™
PA5118920 |
200 μL |
€ 485.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17723134
|
Invitrogen™
PA5121734 |
20 μL |
€ 184.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NT5M Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Dephosphorylates specifically the 5' and 2'(3')- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.Specifications
| NT5M | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| NT5M | |
| 2010013E09Rik; 5' nucleotidase, mitochondrial; 5(3)-deoxyribonucleotidase; 5'(3')-deoxyribonucleotidase, mitochondrial; 5',3'-deoxyribonucleotidase, mitochondrial; 5',3'-nucleotidase, mitochondrial; AI846937; Deoxy-5'-nucleotidase 2; DNT2; dNT-2; mdN; mitochondrial 5' nucleotidase; NT5M | |
| NT5M | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8VCE6, Q9NPB1 | |
| 103850, 56953 | |
| Recombinant protein 5'-Nucleotidase, Mitochondrial. The antigen corresponds to amino acid range 38-228 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title