Learn More
Invitrogen™ NTAL Monoclonal Antibody (NAP-07)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA119329
Product Code. 11524612
  • € 462.00 / 100µg
Description
This antibody reacts with an intracellular epitope of Non-T cell activation linker (NTAL), also known as LAB (linker of activated B cells), a 25 - 30 kDa transmembrane adaptor protein associating with lipid raft components in the plasma membrane of B lymphocytes, NK cells and myeloid cells. Western Blot: Incubation 60 min; non-reducing conditions, 12% separating SDS-PAGE gel.
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NTAL
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
Q9GZY6, Q9JHL0
LAT2
Recombinant cytoplasmic domain (aa 91-243) of human NTAL.
100 μg
Primary
Human, Mouse
Antibody
IgG1
Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
NAP-07
Unconjugated
LAT2
AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5
Mouse
Protein A
RUO
56743, 7462
4°C, do not freeze